Assay Details
| Gene Symbol: Duoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835805.2 | XP_004835862.1 | 5 - 6 | 919 | 82 | |
| XM_021261695.1 | XP_021117354.1 | 5 - 6 | 1387 | 82 | ||
More Information
Set Membership: |
|

Duoxa2
Genomic Map