Assay Details
| Gene Symbol: LOC101701161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874640.2 | XP_004874697.1 | 6 - 7 | 920 | 70 | |
| XM_021252074.1 | XP_021107733.1 | 6 - 7 | 959 | 70 | ||
More Information
Set Membership: |
|

LOC101701161
Genomic Map