Assay Details
| Gene Symbol: Tbcel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870917.2 | XP_004870974.1 | 6 - 7 | 1058 | 73 | |
| XM_004870919.2 | XP_004870976.1 | 6 - 7 | 862 | 73 | ||
| XM_004870920.3 | XP_004870977.1 | 6 - 7 | 868 | 73 | ||
More Information
Set Membership: |
|

Tbcel
Genomic Map