Assay Details
| Gene Symbol: Procr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867878.2 | XP_004867935.1 | 1 - 2 | 258 | 105 | |
| XM_013066736.2 | XP_012922190.1 | 1 - 2 | 258 | 105 | ||
| XM_021246891.1 | XP_021102550.1 | 1 - 2 | 258 | 105 | ||
| XM_021246892.1 | XP_021102551.1 | 2 - 3 | 260 | 105 | ||
| XM_021246893.1 | XP_021102552.1 | 1 - 2 | 258 | 105 | ||
| XM_021246894.1 | XP_021102553.1 | 1 - 2 | 258 | 105 | ||
| XM_021246895.1 | XP_021102554.1 | 1 - 2 | 258 | 105 | ||
More Information
Set Membership: |
|

Procr
Genomic Map