Assay Details
| Gene Symbol: Adcyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853181.3 | XP_004853238.1 | 2 - 3 | 475 | 67 | |
| XM_004853182.3 | XP_004853239.1 | 3 - 4 | 425 | 67 | ||
| XM_021265190.1 | XP_021120849.1 | 3 - 4 | 425 | 67 | ||
More Information
Set Membership: |
|

Adcyap1
Genomic Map