Assay Details
| Gene Symbol: Syt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863422.2 | XP_004863479.1 | 1 - 2 | 259 | 61 | |
| XM_021243269.1 | XP_021098928.1 | 1 - 2 | 262 | 61 | ||
More Information
Set Membership: |
|

Syt9
Genomic Map