Assay Details
| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855740.3 | XP_004855797.1 | 5 - 6 | 607 | 89 | |
| XM_021237165.1 | XP_021092824.1 | 5 - 6 | 611 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myzap| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855740.3 | XP_004855797.1 | 5 - 6 | 607 | 89 | |
| XM_021237165.1 | XP_021092824.1 | 5 - 6 | 611 | 89 | ||