Assay Details
| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855740.3 | XP_004855797.1 | 6 - 7 | 756 | 76 | |
| XM_021237164.1 | XP_021092823.1 | 5 - 6 | 643 | 76 | ||
| XM_021237165.1 | XP_021092824.1 | 6 - 7 | 760 | 76 | ||

Myzap
Genomic Map