Assay Details
| Gene Symbol: Pin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865470.3 | XP_004865527.1 | 1 - 2 | 197 | 64 | |
| XM_004865471.3 | XP_004865528.1 | 1 - 2 | 118 | 64 | ||
More Information
Set Membership: |
|

Pin1
Genomic Map