Assay Details
| Gene Symbol: Wwox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842811.2 | XP_004842868.1 | 1 - 2 | 279 | 66 | |
| XM_021256680.1 | XP_021112339.1 | 1 - 2 | 282 | 66 | ||
| XM_021256681.1 | XP_021112340.1 | 1 - 2 | 264 | 66 | ||
More Information
Set Membership: |
|

Wwox
Genomic Map