Assay Details
| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862714.2 | XP_004862771.1 | 2 - 3 | 490 | 67 | |
| XM_021242869.1 | XP_021098528.1 | 2 - 3 | 527 | 67 | ||
More Information
Set Membership: |
|

Asb4
Genomic Map