Assay Details
| Gene Symbol: Gchfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861579.3 | XP_004861636.1 | 2 - 3 | 244 | 64 | |
| XM_021241931.1 | XP_021097590.1 | 2 - 3 | 196 | 64 | ||
More Information
Set Membership: |
|

Gchfr
Genomic Map