Assay Details
| Gene Symbol: Rdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837343.2 | XP_004837400.1 | 2 - 3 | 360 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh12| Gene Symbol: Rdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837343.2 | XP_004837400.1 | 2 - 3 | 360 | 64 | |
Set Membership: |
|