Assay Details
| Gene Symbol: Ccno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848517.2 | XP_004848574.1 | 1 - 2 | 613 | 104 | |
| XM_021261421.1 | XP_021117080.1 | 2 - 3 | 418 | 104 | ||
More Information
Set Membership: |
|

Ccno
Genomic Map