Assay Details
| Gene Symbol: Rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863039.2 | XP_004863096.1 | 6 - 7 | 572 | 65 | |
| XM_021243020.1 | XP_021098679.1 | 5 - 6 | 515 | 65 | ||
More Information
Set Membership: |
|

Rhebl1
Genomic Map