Assay Details
| Gene Symbol: Abhd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021244154.1 | XP_021099813.1 | 6 - 7 | 793 | 62 | |
| XM_021244155.1 | XP_021099814.1 | 7 - 8 | 829 | 62 | ||
| XM_021244156.1 | XP_021099815.1 | 6 - 7 | 749 | 62 | ||
More Information
Set Membership: |
|

Abhd6
Genomic Map