Assay Details
| Gene Symbol: LOC101723404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858142.2 | XP_004858199.1 | 3 - 4 | 573 | 63 | |
| XM_004858143.2 | XP_004858200.1 | 3 - 4 | 572 | 63 | ||
| XM_013077159.2 | XP_012932613.1 | 3 - 4 | 567 | 63 | ||
| XM_021239002.1 | XP_021094661.1 | 2 - 3 | 426 | 63 | ||
More Information
Set Membership: |
|

LOC101723404
Genomic Map