Assay Details
| Gene Symbol: LOC101723404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858142.2 | XP_004858199.1 | 5 - 6 | 927 | 76 | |
| XM_004858143.2 | XP_004858200.1 | 5 - 6 | 826 | 76 | ||
More Information
Set Membership: |
|

LOC101723404
Genomic Map