Assay Details
| Gene Symbol: CUNH18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855086.2 | XP_004855143.1 | 2 - 3 | 1327 | 86 | |
| XM_004855087.3 | XP_004855144.1 | 2 - 3 | 1553 | 86 | ||
| XM_004855088.3 | XP_004855145.1 | 2 - 3 | 801 | 86 | ||
| XM_021236591.1 | XP_021092250.1 | 2 - 3 | 914 | 86 | ||

CUNH18orf25
Genomic Map