Assay Details
| Gene Symbol: CUNH18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855086.2 | XP_004855143.1 | 4 - 5 | 1632 | 82 | |
| XM_004855087.3 | XP_004855144.1 | 4 - 5 | 1858 | 82 | ||
| XM_004855088.3 | XP_004855145.1 | 4 - 5 | 1106 | 82 | ||
| XM_004855089.2 | XP_004855146.1 | 3 - 4 | 1445 | 82 | ||
| XM_021236591.1 | XP_021092250.1 | 4 - 5 | 1219 | 82 | ||
More Information
Set Membership: |
|

CUNH18orf25
Genomic Map