Assay Details
| Gene Symbol: CUNH5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836223.3 | XP_004836280.1 | 1 - 2 | 220 | 75 | |
| XM_004836224.3 | XP_004836281.1 | 1 - 2 | 217 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836223.3, XM_004836224.3 transcripts.
Set Membership: |
|

CUNH5orf24
Genomic Map