Assay Details
| Gene Symbol: LOC101721083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842876.3 | XP_004842933.1 | 5 - 6 | 531 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101721083| Gene Symbol: LOC101721083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842876.3 | XP_004842933.1 | 5 - 6 | 531 | 80 | |
Set Membership: |
|