Assay Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870002.3 | XP_004870059.1 | 7 - 8 | 849 | 88 | |
XM_013067536.2 | XP_012922990.1 | 7 - 8 | 873 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
