Assay Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842340.2 | XP_004842397.1 | 2 - 3 | 426 | 82 | |
| XM_004842341.3 | XP_004842398.1 | 2 - 3 | 394 | 82 | ||
More Information
Set Membership: |
|

Rcan1
Genomic Map