Assay Details
| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847183.3 | XP_004847240.2 | 9 - 10 | 1206 | 71 | |
| XM_004847184.3 | XP_004847241.2 | 8 - 9 | 1166 | 71 | ||
| XM_013073133.2 | XP_012928587.1 | 10 - 11 | 1382 | 71 | ||
| XM_013073134.2 | XP_012928588.1 | 9 - 10 | 1342 | 71 | ||
More Information
Set Membership: |
|

Rbpj
Genomic Map