Assay Details
| Gene Symbol: Mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859781.2 | XP_004859838.1 | 2 - 3 | 336 | 80 | |
| XM_021239971.1 | XP_021095630.1 | 2 - 3 | 346 | 80 | ||
More Information
Set Membership: |
|

Mmd
Genomic Map