Assay Details
| Gene Symbol: Plppr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869104.2 | XP_004869161.1 | 3 - 4 | 711 | 69 | |
| XM_021247842.1 | XP_021103501.1 | 3 - 4 | 711 | 69 | ||
More Information
Set Membership: |
|

Plppr5
Genomic Map