Assay Details
| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857043.3 | XP_004857100.1 | 5 - 6 | 806 | 66 | |
| XM_004857044.3 | XP_004857101.1 | 5 - 6 | 759 | 66 | ||
| XM_021238296.1 | XP_021093955.1 | 5 - 6 | 719 | 66 | ||
More Information
Set Membership: |
|

Pipox
Genomic Map