Assay Details
| Gene Symbol: Plau | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846313.3 | XP_004846370.1 | 8 - 9 | 1002 | 50 | |
| XM_021259639.1 | XP_021115298.1 | 8 - 9 | 1018 | 50 | ||
More Information
Set Membership: |
|

Plau
Genomic Map