Assay Details
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862431.3 | XP_004862488.1 | 2 - 3 | 331 | 67 | |
| XM_004862432.3 | XP_004862489.1 | 2 - 3 | 331 | 67 | ||
| XM_021242617.1 | XP_021098276.1 | 2 - 3 | 522 | 67 | ||
| XM_021242618.1 | XP_021098277.1 | 2 - 3 | 522 | 67 | ||
More Information
Set Membership: |
|

Ubqln1
Genomic Map