Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861002.3 | XP_004861059.1 | 2 - 3 | 232 | 55 | |
| XM_013078104.2 | XP_012933558.1 | 2 - 3 | 241 | 55 | ||
More Information
Set Membership: |
|

Dcxr
Genomic Map