Assay Details
| Gene Symbol: CUNH1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853583.3 | XP_004853640.3 | 1 - 2 | 239 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH1orf100| Gene Symbol: CUNH1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853583.3 | XP_004853640.3 | 1 - 2 | 239 | 75 | |
Set Membership: |
|