Assay Details
| Gene Symbol: Klf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867690.3 | XP_004867747.1 | 4 - 5 | 954 | 79 | |
| XM_004867691.3 | XP_004867748.1 | 4 - 5 | 849 | 79 | ||
| XM_004867693.3 | XP_004867750.1 | 4 - 5 | 802 | 79 | ||
| XM_021246697.1 | XP_021102356.1 | 4 - 5 | 1022 | 79 | ||
| XM_021246698.1 | XP_021102357.1 | 3 - 4 | 846 | 79 | ||
More Information
Set Membership: |
|

Klf3
Genomic Map