Assay Details
| Gene Symbol: Tctex1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834694.3 | XP_004834751.2 | 2 - 3 | 437 | 77 | |
| XM_004834695.3 | XP_004834752.1 | 2 - 3 | 694 | 77 | ||
More Information
Set Membership: |
|

Tctex1d2
Genomic Map