Assay Details
| Gene Symbol: Mia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078883.2 | XP_012934337.2 | 17 - 18 | 3004 | 134 | |
| XM_021242838.1 | XP_021098497.1 | 11 - 12 | 1050 | 134 | ||
| XM_021242839.1 | XP_021098498.1 | 11 - 12 | 1050 | 134 | ||
More Information
Set Membership: |
|

Mia2
Genomic Map