Assay Details
| Gene Symbol: Mia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078883.2 | XP_012934337.2 | 6 - 7 | 2082 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mia2| Gene Symbol: Mia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078883.2 | XP_012934337.2 | 6 - 7 | 2082 | 83 | |
Set Membership: |
|