Assay Details
| Gene Symbol: Mia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078883.2 | XP_012934337.2 | 8 - 9 | 2234 | 77 | |
| XM_021242838.1 | XP_021098497.1 | 3 - 4 | 307 | 77 | ||
| XM_021242839.1 | XP_021098498.1 | 3 - 4 | 307 | 77 | ||
More Information
Set Membership: |
|

Mia2
Genomic Map