Assay Details
| Gene Symbol: CUNH8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837649.2 | XP_004837706.1 | 1 - 2 | 165 | 104 | |
| XM_004837650.2 | XP_004837707.1 | 1 - 2 | 164 | 104 | ||
| XM_004837651.2 | XP_004837708.1 | 1 - 2 | 165 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004837649.2, XM_004837650.2, XM_004837651.2 transcripts.
Set Membership: |
|

CUNH8orf33
Genomic Map