Assay Details
| Gene Symbol: CUNH8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837649.2 | XP_004837706.1 | 3 - 4 | 561 | 75 | |
| XM_004837650.2 | XP_004837707.1 | 3 - 4 | 554 | 75 | ||
More Information
Set Membership: |
|

CUNH8orf33
Genomic Map