Assay Details
| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844372.3 | XP_004844429.1 | 6 - 7 | 732 | 61 | |
| XM_004844373.3 | XP_004844430.1 | 5 - 6 | 698 | 61 | ||
| XM_021258020.1 | XP_021113679.1 | 6 - 7 | 635 | 61 | ||
| XM_021258021.1 | XP_021113680.1 | 6 - 7 | 813 | 61 | ||
| XM_021258022.1 | XP_021113681.1 | 5 - 6 | 529 | 61 | ||
More Information
Set Membership: |
|

Lman2l
Genomic Map