Assay Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836002.3 | XP_004836059.1 | 6 - 7 | 648 | 64 | |
| XM_004836003.3 | XP_004836060.1 | 6 - 7 | 620 | 64 | ||
| XM_004836004.3 | XP_004836061.1 | 5 - 6 | 599 | 64 | ||
| XM_004836005.3 | XP_004836062.1 | 5 - 6 | 577 | 64 | ||
| XM_021236350.1 | XP_021092009.1 | 4 - 5 | 521 | 64 | ||
More Information
Set Membership: |
|

Fam168b
Genomic Map