Assay Details
| Gene Symbol: Galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852933.3 | XP_004852990.1 | 9 - 10 | 1980 | 64 | |
| XM_004852934.3 | XP_004852991.1 | 9 - 10 | 1980 | 64 | ||
| XM_021264937.1 | XP_021120596.1 | 9 - 10 | 1624 | 64 | ||
More Information
Set Membership: |
|

Galnt7
Genomic Map