Assay Details
| Gene Symbol: Efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837529.2 | XP_004837586.1 | 4 - 5 | 646 | 97 | |
| XM_004837530.2 | XP_004837587.1 | 4 - 5 | 646 | 97 | ||
| XM_004837531.3 | XP_004837588.1 | 4 - 5 | 646 | 97 | ||
| XM_004837533.3 | XP_004837590.1 | 4 - 5 | 622 | 97 | ||
| XM_013068252.2 | XP_012923706.1 | 4 - 5 | 646 | 97 | ||
More Information
Set Membership: |
|

Efcab1
Genomic Map