Assay Details
| Gene Symbol: Vegfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866091.2 | XP_004866148.1 | 4 - 5 | 1283 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vegfd| Gene Symbol: Vegfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866091.2 | XP_004866148.1 | 4 - 5 | 1283 | 94 | |
Set Membership: |
|