Assay Details
| Gene Symbol: Kctd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843617.3 | XP_004843674.2 | 2 - 3 | 426 | 58 | |
| XM_004843618.3 | XP_004843675.2 | 2 - 3 | 426 | 58 | ||
More Information
Set Membership: |
|

Kctd10
Genomic Map