Assay Details
| Gene Symbol: Emsy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863204.3 | XP_004863261.1 | 9 - 10 | 1493 | 66 | |
| XM_004863205.3 | XP_004863262.1 | 9 - 10 | 1491 | 66 | ||
| XM_004863206.3 | XP_004863263.1 | 8 - 9 | 1451 | 66 | ||
| XM_004863207.3 | XP_004863264.1 | 9 - 10 | 1491 | 66 | ||
| XM_004863208.3 | XP_004863265.1 | 8 - 9 | 1447 | 66 | ||
| XM_004863209.3 | XP_004863266.1 | 8 - 9 | 1341 | 66 | ||
| XM_013064981.2 | XP_012920435.1 | 8 - 9 | 1449 | 66 | ||
| XM_021243327.1 | XP_021098986.1 | 9 - 10 | 1340 | 66 | ||
| XM_021243329.1 | XP_021098988.1 | 9 - 10 | 1493 | 66 | ||
| XM_021243330.1 | XP_021098989.1 | 7 - 8 | 1376 | 66 | ||
| XM_021243331.1 | XP_021098990.1 | 8 - 9 | 1340 | 66 | ||
More Information
Set Membership: |
|

Emsy
Genomic Map