Assay Details
| Gene Symbol: Kcnj15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256248.1 | XP_021111907.1 | 3 - 4 | 517 | 81 | |
| XM_021256249.1 | XP_021111908.1 | 4 - 5 | 477 | 81 | ||
| XM_021256250.1 | XP_021111909.1 | 4 - 5 | 424 | 81 | ||
| XM_021256251.1 | XP_021111910.1 | 4 - 5 | 4320 | 81 | ||
| XM_021256252.1 | XP_021111911.1 | 4 - 5 | 386 | 81 | ||
| XM_021256253.1 | XP_021111912.1 | 5 - 6 | 650 | 81 | ||
| XM_021256254.1 | XP_021111913.1 | 4 - 5 | 553 | 81 | ||
| XM_021256255.1 | XP_021111914.1 | 4 - 5 | 332 | 81 | ||
| XM_021256257.1 | XP_021111916.1 | 3 - 4 | 361 | 81 | ||
| XM_021256258.1 | XP_021111917.1 | 2 - 3 | 120 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021256248.1, XM_021256249.1, XM_021256250.1, XM_021256251.1, XM_021256252.1, XM_021256253.1, XM_021256254.1, XM_021256255.1, XM_021256257.1, XM_021256258.1 transcripts.
Set Membership: |
|

Kcnj15
Genomic Map