Assay Details
| Gene Symbol: Decr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873771.3 | XP_004873828.1 | 1 - 2 | 243 | 69 | |
| XM_021251428.1 | XP_021107087.1 | 2 - 3 | 939 | 69 | ||
| XM_021251429.1 | XP_021107088.1 | 2 - 3 | 961 | 69 | ||
| XM_021251430.1 | XP_021107089.1 | 1 - 2 | 1068 | 69 | ||
| XM_021251431.1 | XP_021107090.1 | 1 - 2 | 1068 | 69 | ||
More Information
Set Membership: |
|

Decr2
Genomic Map