Assay Details
Gene Symbol: CUNH11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852011.3 | XP_004852068.1 | 6 - 7 | 655 | 72 | |
XM_004852012.3 | XP_004852069.1 | 5 - 6 | 551 | 72 | ||
XM_021264333.1 | XP_021119992.1 | 6 - 7 | 582 | 72 | ||
XM_021264334.1 | XP_021119993.1 | 6 - 7 | 546 | 72 |
