Assay Details
Gene Symbol: CUNH11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852011.3 | XP_004852068.1 | 7 - 8 | 793 | 76 | |
XM_004852012.3 | XP_004852069.1 | 6 - 7 | 689 | 76 | ||
XM_021264333.1 | XP_021119992.1 | 7 - 8 | 720 | 76 | ||
XM_021264334.1 | XP_021119993.1 | 7 - 8 | 684 | 76 | ||
XM_021264335.1 | XP_021119994.1 | 4 - 5 | 499 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
